Merck
  • Influence of environmental hypertonicity on the induction of ureogenesis and amino acid metabolism in air-breathing walking catfish (Clarias batrachus, Bloch).
Influence of environmental hypertonicity on the induction of ureogenesis and amino acid metabolism in air-breathing walking catfish (Clarias batrachus, Bloch).
Indian journal of experimental biology (2014-07-26)
Bodhisattwa Banerjee, Gitalee Bhuyan, Nirmalendu Saha
ZUSAMMENFASSUNG
Effect of environmental hypertonicity, due to exposure to 300 mM mannitol solution for 7 days, on the induction of ureogenesis and also on amino acid metabolism was studied in the air-breathing walking catfish, C. batrachus, which is already known to have the capacity to face the problem of osmolarity stress in addition to other environmental stresses in its natural habitats. Exposure to hypertonic mannitol solution led to reduction of ammonia excretion rate by about 2-fold with a concomitant increase of urea-N excretion rate by about 2-fold. This was accompanied by significant increase in the levels of both ammonia and urea in different tissues and also in plasma. Further, the environmental hypertonicity also led to significant accumulation of different non-essential free amino acids (FAAs) and to some extent the essential FAAs, thereby causing a total increase of non-essential FAA pool by 2-3-fold and essential FAA pool by 1.5-2.0-fold in most of the tissues studied including the plasma. The activities of three ornithine-urea cycle (OUC) enzymes such as carbamoyl phosphate synthetase, argininosuccinate synthetase and argininosuccinate lyase in liver and kidney tissues, and four key amino acid metabolism-related enzymes such as glutamine synthetase, glutamate dehydrogenase (reductive amination), alanine aminotransaminase and aspartate aminotransaminase were also significantly up-regulated in different tissues of the fish while exposing to hypertonic environment. Thus, more accumulation and excretion of urea-N observed during hypertonic exposure were probably associated with the induction of ureogenesis through the induced OUC, and the increase of amino acid pool was probably mainly associated with the up-regulation of amino acid synthesizing machineries in this catfish in hypertonic environment. These might have helped the walking catfish in defending the osmotic stress and to acclimatize better under hypertonic environment, which is very much uncommon among freshwater teleosts.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ammoniak -Lösung, 7 N in methanol
Sigma-Aldrich
DL-Dithiothreitol -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Harnstoff, powder, BioReagent, Molecular Biology, suitable for cell culture
Supelco
DL-Dithiothreitol -Lösung, 1 M in H2O
Sigma-Aldrich
Harnstoff, ACS reagent, 99.0-100.5%
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Supelco
Harnstoff, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Harnstoff -Lösung, BioUltra, ~8 M in H2O
Supelco
D-Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
Ammoniak -Lösung, 2.0 M in ethanol
Sigma-Aldrich
Harnstoff, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
Sigma-Aldrich
Harnstoff, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Ammoniak, anhydrous, ≥99.98%
Sigma-Aldrich
Harnstoff, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Ammoniak -Lösung, 0.4 M in dioxane
Sigma-Aldrich
Ammoniak -Lösung, 4 M in methanol
USP
Mannitol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
L-Ornithin -monohydrochlorid, ≥99%
Sigma-Aldrich
Ammoniak -Lösung, 0.4 M in THF
Sigma-Aldrich
Harnstoff, suitable for electrophoresis
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
Ammoniak -Lösung, 2.0 M in isopropanol
Sigma-Aldrich
Harnstoff, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
L-Ornithin -monohydrochlorid, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Supelco
Harnstoff, analytical standard
Mannitol, European Pharmacopoeia (EP) Reference Standard