- Safety profile of extended-release niacin in the AIM-HIGH trial.
Safety profile of extended-release niacin in the AIM-HIGH trial.
The New England journal of medicine (2014-07-12)
Todd J Anderson, William E Boden, Patrice Desvigne-Nickens, Jerome L Fleg, Moti L Kashyap, Ruth McBride, Jeffrey L Probstfield
PMID25014706
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Nicotinsäure, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Nicotinsäure, European Pharmacopoeia (EP) Reference Standard