Merck
  • Effects of chemically induced hypoxia on in vitro expression of hypoxia inducible factor-lα, vascular endothelial growth factor, aggrecanase-1, and tissue inhibitor of metalloproteinase-3 in rat mandibular condylar chondrocytes.
Effects of chemically induced hypoxia on in vitro expression of hypoxia inducible factor-lα, vascular endothelial growth factor, aggrecanase-1, and tissue inhibitor of metalloproteinase-3 in rat mandibular condylar chondrocytes.
Journal of oral & facial pain and headache (2014-07-30)
Huiping Huang, Jia Yu, Dahai Yu, Yu Li, Shujian Lu
ZUSAMMENFASSUNG
To examine the expression of hypoxia inducible factor-lα (HIF-1α), vascular endothelial growth factor (VEGF), aggrecanase-1 (ADAMTS-4), and tissue inhibitor of metalloproteinase-3 (TIMP-3) in rat mandibular condylar chondrocytes under hypoxic conditions and examine the relationship of these expressed factors in early condylar cartilage growth. CoCl2 was used to mimic a hypoxic microenvironment by chemically inducing hypoxia. Chondrocytes, which were obtained from the mandibular condyles of 3-week-old female Sprague-Dawley rats, were treated with 125 μmol/L CoCl2 for 48 hours. The HIF-1α, VEGF, ADAMTS-4, and TIMP-3 mRNA levels in chondrocytes were detected using a semiquantitativepolymerase chain reaction (PCR) at 12, 24, and 48 hours after the initiation of hypoxia and normoxia conditions. A univariate variance analysis (pairwise least significant difference t test) using a SPSS 13.0 software package was performed to test for differences between different groups. PCR analysis of the chondrocytes showed that the expression of HIF-1α and VEGF mRNA increased at 12, 24, and 48 hours after induction of hypoxia. HIF-1α expression gradually increased throughout the study duration, while VEGF expression peaked at 12 hours. Compared to normoxia conditions, hypoxia resulted in constantly elevated expression levels of both. On the other hand, there was no significant difference in the ADAMTS-4 and TIMP-3 mRNA expression between hypoxic and normoxic conditions throughout the study (P > .05). An upregulated HIF-1α and VEGF mRNA expression indicates their important roles in cartilage vascularization and development in newly hypoxic microenvironments. Additionally, chemically induced hypoxia has little effect on the expressions of ADAMTS-4 and TIMP-3.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cobalt(II)-chlorid, 97%
Sigma-Aldrich
Cobalt(II)-chlorid, purum p.a., anhydrous, ≥98.0% (KT)
Sigma-Aldrich
Cobaltchlorid 0,1 mol/l -Lösung
Sigma-Aldrich
Kobalt, powder, 2 μm particle size, 99.8% trace metals basis
Sigma-Aldrich
Kobalt, powder, <150 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Cobalt(II)-chlorid, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Kobalt, granular, 99.99% trace metals basis
Sigma-Aldrich
Kobalt, Carbon coated magnetic, nanopowder, <50 nm particle size (TEM), ≥99%
Sigma-Aldrich
Cobalt(II)-chlorid, AnhydroBeads, −10 mesh, 99.995% trace metals basis
Sigma-Aldrich
Kobalt, pieces, 99.5% trace metals basis
Sigma-Aldrich
Kobalt, wire, diam. 1.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Kobalt, foil, thickness 0.1 mm, 99.95% trace metals basis
Sigma-Aldrich
Kobalt, foil, thickness 1.0 mm, 99.95% trace metals basis
Sigma-Aldrich
Kobalt, foil, thickness 0.1 mm, ≥99.99%
Sigma-Aldrich
Kobalt, rod, diam. 5.0 mm, 99.95% trace metals basis
Kobalt, foil, light tested, 25x25mm, thickness 0.01mm, 99.9%
Kobalt, foil, 4mm disks, thickness 0.05mm, annealed, 99.9%
Kobalt, foil, 8mm disks, thickness 0.003mm, 99.9%
Kobalt, foil, 6mm disks, thickness 0.01mm, 99.9%
Kobalt, foil, 4mm disks, thickness 0.007mm, 99.9%
Kobalt, foil, 8mm disks, thickness 0.01mm, 99.9%
Kobalt, foil, 25x25mm, thickness 0.25mm, as rolled, 99.9%
Kobalt, foil, light tested, 25x25mm, thickness 0.05mm, as rolled, 99.99+%
Kobalt, foil, light tested, 50x50mm, thickness 0.0125mm, 99.9%
Kobalt, wire reel, 0.5m, diameter 0.05mm, hard, 99.9%
Kobalt, foil, 25mm disks, thickness 0.125mm, annealed, 99.9%
Kobalt, foil, 8mm disks, thickness 0.006mm, 99.9%
Kobalt, foil, 4mm disks, thickness 0.01mm, 99.9%
Kobalt, foil, 4mm disks, thickness 0.025mm, 99.9%
Kobalt, foil, 6mm disks, thickness 0.008mm, 99.9%