Merck
  • Solid-phase extraction of copper(II) in water and food samples using silica gel modified with bis(3-aminopropyl)amine and determination by flame atomic absorption spectrometry.
Solid-phase extraction of copper(II) in water and food samples using silica gel modified with bis(3-aminopropyl)amine and determination by flame atomic absorption spectrometry.
Journal of AOAC International (2014-08-26)
Duygu Cagirdi, Hüseyin Altundag, Mustafa Imamoglu, Mustafa Tuzen
ZUSAMMENFASSUNG
A simple and selective separation and preconcentration method was developed for the determination of Cu(ll) ions. This method is based on adsorption of Cu(ll) ions from aqueous solution on a bis(3-aminopropyl)amine modified silica gel column and flame atomic absorption spectrometric determination after desorption. Various analytical parameters such as pH, type of eluent solution and its volume, flow rate of sample and eluent, and sample volume were optimized. Effects of some cation, anion, and transition metal ions on the recoveries of Cu(ll) ions were also investigated. Cu(ll) ions were quantitatively recovered at pH 6; 5.0 mL of 2 M HCI was used as the eluent. The preconcentration factor was found to be 150. The LOD was 0.12 microg/L for Cu(ll). The accuracy of the method was confirmed by analysis of Tea Leaves (INCT-TL-1) and Fish Protein (DORM-3) certified reference materials. The optimized method was applied to various water and food samples for the determination of Cu(ll).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
LUDOX® AS-40 kolloidales Siliziumdioxid, 40 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® HS-40 kolloidales Siliziumdioxid, 40 wt. % suspension in H2O
Sigma-Aldrich
Siliciumdioxid, nanopowder, 10-20 nm particle size (BET), 99.5% trace metals basis
Sigma-Aldrich
LUDOX® TM-50 kolloidales Siliziumdioxid, 50 wt. % suspension in H2O
Sigma-Aldrich
Kupfer, powder, <425 μm, 99.5% trace metals basis
Sigma-Aldrich
Kupfer, foil, thickness 0.25 mm, 99.98% trace metals basis
Sigma-Aldrich
Kupfer, powder, 99.99% trace metals basis
Sigma-Aldrich
Silicium(IV)-oxid, nanoparticles, mesoporous, 200 nm particle size, pore size 4 nm
Sigma-Aldrich
Siliciumdioxid, nanopowder (spherical, porous), 5-20 nm particle size (TEM), 99.5% trace metals basis
Sigma-Aldrich
Silicium(IV)-oxid, nanopowder, 99.8% trace metals basis
Sigma-Aldrich
LUDOX® HS-30 kolloidales Siliziumdioxid, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® SM kolloidales Siliziumdioxid, 30 wt. % suspension in H2O
Sigma-Aldrich
Siliziumdioxid, mesostrukturiert, MCM-41 type (hexagonal)
Sigma-Aldrich
LUDOX® TM-40 kolloidales Siliziumdioxid, 40 wt. % suspension in H2O
Sigma-Aldrich
Silicium(IV)-oxid
Sigma-Aldrich
Kupfer, powder, <75 μm, 99%
Sigma-Aldrich
Kupfer, wire, diam. 1.0 mm, ≥99.9%
Sigma-Aldrich
LUDOX® AS-30 kolloidales Siliziumdioxid, 30 wt. % suspension in H2O
Sigma-Aldrich
Kupfer, nanopowder, 40-60 nm particle size (SAXS), ≥99.5% trace metals basis
Sigma-Aldrich
Kupfer, powder (spheroidal), 10-25 μm, 98%
Sigma-Aldrich
LUDOX® AM kolloidales Siliziumdioxid, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® TMA kolloidales Siliziumdioxid, 34 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® CL kolloidales Siliziumdioxid, 30 wt. % suspension in H2O
Sigma-Aldrich
Kupfer, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
Kupfer, turnings, purum p.a., ≥99.0%
Sigma-Aldrich
Kupfer, nanopowder, 60-80 nm particle size (APS), ≥99.5% trace metals basis
Sigma-Aldrich
LUDOX® LS kolloidales Siliziumdioxid, 30 wt. % suspension in H2O
Sigma-Aldrich
Kupfer, ACS reagent, granular, 10-40 mesh, ≥99.90%
Sigma-Aldrich
Kupfer, beads, 2-8 mm, 99.9995% trace metals basis
Sigma-Aldrich
Kupfer, foil, ≥99.8% (complexometric)