Merck
  • A review of methods for the analysis of orphan and difficult pesticides: glyphosate, glufosinate, quaternary ammonium and phenoxy acid herbicides, and dithiocarbamate and phthalimide fungicides.
A review of methods for the analysis of orphan and difficult pesticides: glyphosate, glufosinate, quaternary ammonium and phenoxy acid herbicides, and dithiocarbamate and phthalimide fungicides.
Journal of AOAC International (2014-08-26)
Renata Raina-Fulton
ZUSAMMENFASSUNG
This article reviews the chromatography/MS methodologies for analysis of pesticide residues of orphan and difficult chemical classes in a variety of sample matrixes including water, urine, blood, and food. The review focuses on pesticide classes that are not commonly included in multiresidue analysis methods such as highly polar or ionic herbicides including glyphosate, glufosinate, quaternary ammonium, and phenoxy acid herbicides, and some of their major degradation or metabolite products. In addition, dithiocarbamate and phthalimide fungicides, which are thermally unstable and have stability issues in some solvents or sample matrixes, are also examined due to their special needs in residue analysis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glycin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycin, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycin, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycin -hydrochlorid, ≥99% (HPLC)
Supelco
Glyphosat, PESTANAL®, analytical standard
Supelco
Glufosinatammonium, PESTANAL®, analytical standard
SAFC
Glycin
Supelco
Glycin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phthalimid, ≥99%
Sigma-Aldrich
Glycin, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycin, ACS reagent, ≥98.5%
Sigma-Aldrich
N-(Phosphonomethyl)glycin, 96%
Sigma-Aldrich
Phthalimid-Kalium, 98%
Sigma-Aldrich
Glycin, 99%, FCC
USP
Glycin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Glycin, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycin, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Glycin 1 M -Lösung
Sigma-Aldrich
N-(Phosphonomethyl)glycin, BioReagent, suitable for plant cell culture
Supelco
Glycin, analytical standard, for nitrogen determination according to Kjeldahl method
Glycin, European Pharmacopoeia (EP) Reference Standard
Supelco
Glycin -Lösung, 100 mM amino acid in 0.1 M HCl, analytical standard
Sigma-Aldrich
Phthalimid-Kalium, purum, ≥99.0% (NT)
Supelco
Glycin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Glycin, tested according to Ph. Eur.