Merck
  • An effective utilization of the slag from acid leaching of coal-waste: preparation of water glass with a low-temperature co-melting reaction.
An effective utilization of the slag from acid leaching of coal-waste: preparation of water glass with a low-temperature co-melting reaction.
Journal of the Air & Waste Management Association (1995) (2014-09-05)
Li Fang, Xiaofang Duan, Rongming Chen, Fangqin Cheng
ZUSAMMENFASSUNG
This paper presents an effective utilization of slag from acid leaching of coal-waste with a novel approach, namely low-temperature co-melting method, for preparation of sodium silicate (Na2O x nSiO2) using slag from acid leaching of coal-waste as feedstock. It is very interesting that the co-melting reaction temperature of the mixture of Na2CO3 and the feedstock (50-100 microm) was as low as 850 degrees C, which was significantly lower than the temperature used in traditional sodium silicate production (1400 degrees C). The optimum SiO2/Na2O ratio was identified as 7:3 according to the results of thermogravimetry-differential scanning calorimetry (TGA-DSC), ICP-AES, and X-ray diffraction (XRD) analyses. In this condition, the main product was sodium disilicate (Na2O x 2SiO2), with water solubility of 85.0%. More importantly, the impurities such as aluminum in the feedstock, which had adverse effect on subsequent treatment, were concentrated almost completely in the filter residue as insoluble sodium alumunosilicates, i.e., Na(Si2Al)O6 x H2O. The lower co-melting temperature of this process demonstrates a significant energy-saving opportunity and thus a promising approach for highly effective utilization of coal-waste. Implications: Recently, alumina extraction from coal-waste has been extensively investigated and industrial applied in China. However, the slag-containing silica generated from the acid leaching process of coal-waste led to a secondary pollution, which hindered large-scale production. The proposed low-temperature co-melting method for preparation of sodium silicate (Na2O x nSiO2) using slag from acid leaching of coal-waste as feedstock indicated that it is an efficient approach for the recovery of silica from the acid-leached slag of coal-waste with minimal environmental impact.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumcarbonat, powder, ≥99.5%, ACS reagent
Sigma-Aldrich
Natriumcarbonat, ACS reagent, anhydrous, ≥99.5%, powder or granules
Sigma-Aldrich
Natriumcarbonat, ACS reagent (primary standard), anhydrous, 99.95-100.05% dry basis
Sigma-Aldrich
Natriumcarbonat, ReagentPlus®, ≥99.5%
Sigma-Aldrich
LUDOX® AS-40 kolloidales Siliziumdioxid, 40 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® HS-40 kolloidales Siliziumdioxid, 40 wt. % suspension in H2O
Sigma-Aldrich
Siliciumdioxid, nanopowder, 10-20 nm particle size (BET), 99.5% trace metals basis
Sigma-Aldrich
Natriumcarbonat, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Natriumcarbonat, BioXtra, ≥99.0%
Sigma-Aldrich
LUDOX® TM-50 kolloidales Siliziumdioxid, 50 wt. % suspension in H2O
Sigma-Aldrich
Natriumcarbonat, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Natriumoxid, 80%
Sigma-Aldrich
Silicium(IV)-oxid, nanoparticles, mesoporous, 200 nm particle size, pore size 4 nm
Sigma-Aldrich
Siliciumdioxid, nanopowder (spherical, porous), 5-20 nm particle size (TEM), 99.5% trace metals basis
Supelco
Natriumcarbonat-Konzentrat, Na2CO3 72 mM in water, IC eluent concentrate (20x) for Metrosep A Supp 7
Sigma-Aldrich
LUDOX® HS-30 kolloidales Siliziumdioxid, 30 wt. % suspension in H2O
Sigma-Aldrich
Silicium(IV)-oxid, nanopowder, 99.8% trace metals basis
Supelco
Natriumcarbonat-Konzentrat, 0.1 M Na2CO3 in water, eluent concentrate for IC
Sigma-Aldrich
LUDOX® SM kolloidales Siliziumdioxid, 30 wt. % suspension in H2O
Sigma-Aldrich
Siliziumdioxid, mesostrukturiert, MCM-41 type (hexagonal)
Sigma-Aldrich
LUDOX® TM-40 kolloidales Siliziumdioxid, 40 wt. % suspension in H2O
Sigma-Aldrich
Silicium(IV)-oxid
Sigma-Aldrich
LUDOX® AS-30 kolloidales Siliziumdioxid, 30 wt. % suspension in H2O
Supelco
Natriumcarbonat, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
LUDOX® AM kolloidales Siliziumdioxid, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® TMA kolloidales Siliziumdioxid, 34 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® CL kolloidales Siliziumdioxid, 30 wt. % suspension in H2O
Sigma-Aldrich
Natriumcarbonat, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Sigma-Aldrich
LUDOX® LS kolloidales Siliziumdioxid, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® CL-X kolloidales Siliziumdioxid, 45 wt. % suspension in H2O