Merck
  • Ultrasonic assisted synthesis, acoustical property and antibacterial activity of some Schiff bases.
Ultrasonic assisted synthesis, acoustical property and antibacterial activity of some Schiff bases.
International Letters of Chemistry, Physics and Astronomy (2014)
Premalatha R and Santhi N.