Merck
Extent and limitation of the control of nuclear apoptosis by DNA-fragmenting factor.
Biochemical and biophysical research communications (1999-01-28)
W Wöhrl, G Häcker
ZUSAMMENFASSUNG
During apoptosis, changes to the nucleus of the dying cell include DNA degradation and structural collapse. These changes are accomplished by caspase-mediated cleavage of DNA-fragmenting factor DFF45, an inhibitor of the effector molecule DFF40. DFF45 and, more efficiently, a mutant lacking one caspase-cleavage site (DFF45m) inhibited nuclear changes in a cell-free system when apoptosis was initiated by adding caspase-3 to cell extracts. In primary tissues from several mammalian species, human caspase-3 activated and human DFF45m blocked nuclear apoptosis demonstrating evolutionary conservation of this step. However, DFF45m did not significantly inhibit DNA-fragmenting activity in extracts from staurosporine-treated cells from the human cell line Jurkat. In extracts from normal Jurkat cells, DFF45m blocked caspase-triggered DNA cleavage efficiently only if added within a short time of the addition of the caspase. At later time points, this inhibition by DFF45m was strongly reduced in efficiency while Zn2+ still completely blocked DNA fragmentation. These results demonstrate the evolutionary conservation of a linear pathway in apoptosis and suggest the existence of more complex events as final effector machinery.