- Tissue localization of nerve growth factor and nerve growth factor receptors.
Tissue localization of nerve growth factor and nerve growth factor receptors.
Current topics in microbiology and immunology (1991-01-01)
M Bothwell
PMID1851699
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Nerve Growth Factor-β human, recombinant, expressed in HEK 293 cells, lyophilized powder, suitable for cell culture