Merck
  • The reaction of ethyl benzoylacetate with malononitrile: a novel synthesis of some pyridazine, pyridazino [2, 3-a] quinazoline and pyrrole derivatives.
The reaction of ethyl benzoylacetate with malononitrile: a novel synthesis of some pyridazine, pyridazino [2, 3-<I>a</I>] quinazoline and pyrrole derivatives.
Tetrahedron (2001)
Abdelrazek FM, et al.