Merck
GPR34 is a receptor for lysophosphatidylserine with a fatty acid at the sn-2 position.
Journal of biochemistry (2012-02-22)
Hajime Kitamura, Kumiko Makide, Akira Shuto, Masaya Ikubo, Asuka Inoue, Kensuke Suzuki, Yusuke Sato, Sho Nakamura, Yuko Otani, Tomohiko Ohwada, Junken Aoki
ZUSAMMENFASSUNG
GPR34 is a G protein-coupled receptor belonging to the P2Y family. Here, we attempted to resolve conflicting reports about whether it is a functional lysophosphatidylserine (LysoPS) receptor. In HEK293 cells expressing human, mouse or rat GPR34 and Gα chimera between Gαq and Gαi1(Gq/i1), LysoPS quickly elevated intracellular Ca(2+) ion levels ([Ca(2+)](i)). LysoPS also stimulated alkaline phosphatase (AP)-tagged TGFα (AP-TGFα) release in GPR34-expressing HEK293 cells and induced the migration of CHO-K1 cells expressing GPR34. Other lysophospholipids did not induce these actions. Replacement of the serine residue of LysoPS abolished the reactivity of LysoPS with GPR34, indicating that GPR34 strictly recognizes the serine head group of LysoPS. Recombinant phosphatidylserine-specific phospholipase A(1) (PS-PLA(1)) that deacylates fatty acid at the sn-1 position of PS and produces 2-acyl-LysoPS, but not catalytically inactive mutant PS-PLA(1), stimulated the release of AP-TGFα from GPR34-expressing cells. Consistent with the result, LysoPS was detected in the cells treated with wild-type PS-PLA(1) but not with the mutant PS-PLA(1). PS treated with PLA(1) was much more effective at stimulating AP-TGFα release than PS treated with PLA(2). In addition, migration-resistant 2-acyl-1-deoxy-LysoPS, a 2-acyl-LysoPS analogue, was much more potent than 1-acyl-2-deoxy-LysoPS. The present studies confirm that GPR34 is a cellular receptor for LysoPS, especially with a fatty acid at the sn-2 position.