Merck
  • [A study of quantitative dynamics of F-actin during oocyte maturation in the starfish Asterias amurensis].
[A study of quantitative dynamics of F-actin during oocyte maturation in the starfish Asterias amurensis].
Ontogenez (2006-10-07)
N E Lamash, M G Eliseĭkina
ZUSAMMENFASSUNG
We studied the actin cytoskeleton state in Asterias amurensis oocytes within 30 min after the 1-methyladenine-induced maturation until the germinal vesicle breakdown. The total amount of actin remained unchanged during oocyte maturation. In immature oocytes, the major part of actin is not a part of filaments, but in the presence of 1-methyladenine massive actin polymerization began already within 20 min. Electron immunocytochemistry methods demonstrated joint localization of actin and alpha-protein in the cytoplasm. They were redistributed from the cortex to the cytoplasm in the presence of 1-methyladenine. A possible involvement of actin cytoskeleton in transmembrane transduction of the hormonal signal at the postreceptor stages is discussed.