Merck
  • High expression of APAF-1 elevates erythroid apoptosis in iron overload myelodysplastic syndrome.
High expression of APAF-1 elevates erythroid apoptosis in iron overload myelodysplastic syndrome.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine (2013-10-22)
Shucheng Gu, Youshan Zhao, Juan Guo, Feng Xu, Chengming Fei, Xi Zhang, Chao Xiao, Chunkang Chang, Xiao Li
ZUSAMMENFASSUNG
Apoptotic protease-activating factor 1 (APAF-1) is a central component of the intrinsic pathway of apoptosis. Our study aims at searching the role of APAF-1 in iron overload myelodysplastic syndrome (MDS). Erythroid apoptosis rate, mRNA expression levels of APAF-1, and caspase-9 activity were determined by flow cytometry, quantitative real-time PCR, and colorimetric assay in MDS patients, respectively. In addition, K562 and MDS-L cell lines were incubated with different concentrations of ferric ammonium citrate (FAC) or ferric ammonium citrate + desferrioxamine (FAC + DFO) in vitro to observe the alteration in erythrocyte apoptosis rate, APAF-1 mRNA, and protein expression levels. Moreover, as control, erythroid apoptosis rate and APAF-1 mRNA expression were detected after silencing APAF-1 expression by endoribonuclease-prepared small interfering RNAs (esiRNAs) in K562 and MDS-L cell lines. Both erythroid apoptosis rate and APAF-1 mRNA expression of the iron overload (IO) group were significantly higher than those of the non-IO group (P < 0.001 and P < 0.001). There is a significant difference of caspase-9 activity between the IO group and the non-IO group (P < 0.001). Erythroid apoptosis rate and APAF-1 mRNA expression of K562 and MDS-L cell lines significantly elevated after FAC incubation in different concentrations (P < 0.001 and P < 0.001 for K562; P < 0.001 and P < 0.001 for MDS-L), while erythroid apoptosis rate and APAF-1 mRNA expression in the FAC + DFO group declined (P < 0.001 and P < 0.001 for K562; P < 0.001 and P < 0.001 for MDS-L). After silencing of APAF-1 expression with specific esiRNAs, erythroid apoptosis rate and APAF-1 mRNA expression of K562 and MDS-L cell lines markedly decreased (P < 0.001 and P < 0.001 for K562; P < 0.001 and P < 0.001 for MDS-L). APAF-1 plays an important role in iron-induced erythroid apoptosis increase in MDS.