- Presence of an expressed beta-tubulin gene (TUBB) in the HLA class I region may provide the genetic basis for HLA-linked microtubule dysfunction.
Presence of an expressed beta-tubulin gene (TUBB) in the HLA class I region may provide the genetic basis for HLA-linked microtubule dysfunction.
Human genetics (1994-01-01)
A Volz, E Weiss, J Trowsdale, A Ziegler
PMID8270253
ZUSAMMENFASSUNG
An expressed beta-tubulin gene (TUBB) has previously been localized to chromosome region 6pter-p21 in man. By using a panel of deletion mutant cell lines and radiation-reduced hybrids containing fragments of chromosome 6, the TUBB locus could be mapped to the HLA class I region at 6p21.3. A long range restriction map including TUBB and several HLA class I genes was then generated by rotating field gel electrophoresis. The results show that TUBB maps to a segment 170-370 kb telomeric of HLA-C. This location suggests that a mutation at the TUBB locus could be the cause for certain forms of HLA-linked microtubule dysfunction, including immotile cilia syndrome.