Merck
  • Suppression of immunoglobulin E-mediated allergic responses by regulator of G protein signaling 13.
Suppression of immunoglobulin E-mediated allergic responses by regulator of G protein signaling 13.
Nature immunology (2007-11-21)
Geetanjali Bansal, Zhihui Xie, Sudhir Rao, Karl H Nocka, Kirk M Druey
ZUSAMMENFASSUNG
Mast cells elicit allergic responses through degranulation and release of proinflammatory mediators after antigen crosslinking of the immunoglobulin E receptor FcepsilonRI. Proteins of the 'regulator of G protein signaling' (RGS) family negatively control signaling mediated by G protein-coupled receptors through GTPase-accelerating protein activity. Here we show that RGS13 inhibited allergic responses by physically interacting with the regulatory p85alpha subunit of phosphatidylinositol-3-OH kinase in mast cells and disrupting its association with an FcepsilonRI-activated scaffolding complex. Rgs13-/- mice had enhanced immunoglobulin E-mediated mast cell degranulation and anaphylaxis. Thus, RGS13 inhibits the assembly of immune receptor-induced signalosomes in mast cells. Abnormal RGS13 expression or function may contribute to disorders of amplified mast cell activity, such as idiopathic anaphylaxis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Dinitrophenyl-Antikörper, Maus monoklonal, IgE isotype, ~1 mg/mL, clone SPE-7, affinity purified immunoglobulin, buffered aqueous solution