- Fibronectin at a glance.
Fibronectin at a glance.
Journal of cell science (2002-09-24)
Roumen Pankov, Kenneth M Yamada
PMID12244123
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Fibronectin, recombinant, expressed in HEK 293 cells, lyophilized powder, suitable for cell culture
Roche
Fibronektin (rein), from human plasma, lyophilized (clear, colorless solution after reconstitution)
Sigma-Aldrich
Fibronektin, Rinderplasma, solution, Extracellular matrix (ECM) glycoprotein, Suitable for cell adhesion, growth, migration, and differentiation.