Merck
Expression of beta-synuclein in normal human astrocytes.
Neuroreport (2001-10-06)
K Tanji, F Mori, S Nakajo, T Imaizumi, H Yoshida, T Hirabayashi, M Yoshimoto, K Satoh, H Takahashi, K Wakabayashi
ZUSAMMENFASSUNG
Although alpha- and beta-synucleins are expressed predominantly in presynaptic nerve terminals, recent studies have demonstrated that alpha-synuclein is also expressed in cultured astrocytes and oligodendrocytes. We determined whether beta-synuclein might be expressed in astrocytes. Beta-synuclein mRNA and protein were detected in normal human astrocytes in culture, and immunofluorescent staining showed that beta-synuclein protein was expressed within the cytoplasm and nucleus. Furthermore, beta-synuclein immunoreactivity was present in astrocytes, but not in oligodendrocytes, in normal human brain tissues. Ultrastructurally, beta-synuclein immunoreactivity was found in the cytoplasm of astrocytes, in association with the plasma membrane, ribosomes, rough endoplasmic reticulum and the nuclear outer membrane. The novel expression of beta-synuclein in astrocytes may provide an important insight about the role of this protein.