Merck
  • CaMKK-CaMKI signaling pathways differentially control axon and dendrite elongation in cortical neurons.
CaMKK-CaMKI signaling pathways differentially control axon and dendrite elongation in cortical neurons.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2010-02-26)
April P Neal, Elizabeth Molina-Campos, Brenda Marrero-Rosado, Aaron B Bradford, Sara M Fox, Natalia Kovalova, Heidi E Hannon