Merck
  • Novel 3D, 2D, and 0D first-row coordination compounds with 4,4'-bipyridine-N,N'-dioxide incorporating sulfur-containing anions.
Novel 3D, 2D, and 0D first-row coordination compounds with 4,4'-bipyridine-N,N'-dioxide incorporating sulfur-containing anions.
Inorganic chemistry (2006-04-11)
Déborah González Mantero, Antonia Neels, Helen Stoeckli-Evans
ZUSAMMENFASSUNG
The reaction of M(S2O6) (M = Cu(II), Ni(II), and Co(II)) with 4,4'-bipyridine-N,N'-dioxide (bpdo) results in the formation of novel 3D, 2D, and mononuclear complexes. Complex 1, {[Cu(H2O)(bpdo)2](S2O6)(H2O)}n, is a 2-D wavelike polymer with the Cu(II) ion located on a 2-fold axis and having a distorted square-pyramidal coordination sphere. With Co(II) and Ni(II), 3-D complexes, {[M(bpdo)3](S2O6)(C2H5OH)7}n [M = Co(II) (2), Ni(II) (3)], were obtained. The metal atoms are situated on centers of symmetry and have octahedral environments coordinated to six bpdo molecules. The same reaction in aqueous solution with a metal/ligand ratio of 1:1 results in the formation of mononuclear complexes, {[M(bpdo)(H2O)5](SO4)(H2O)2} [M = Co(II) (4), Ni(II) (5)], accompanied by the decomposition of the dithionate anions S2O6(2-) to sulfate anions SO4(2-).