Merck
Prolidase is required for early trafficking events during influenza A virus entry.
Journal of virology (2014-07-18)
Marie O Pohl, Thomas O Edinger, Silke Stertz
ZUSAMMENFASSUNG
Influenza A virus (IAV) entry is a multistep process that requires the interaction of the virus with numerous host factors. In this study, we demonstrate that prolidase (PEPD) is a cellular factor required by IAV for successful entry into target cells. PEPD was selected as a candidate during an entry screen performed on nonvalidated primary hits from previously published genome-wide small interfering RNA (siRNA) screens. siRNA-mediated depletion of PEPD resulted in the decreased growth of IAV during mono- and multicycle growth. This growth defect was independent of cell type or virus strain. Furthermore, IAV restriction was apparent as early as 3 h postinfection, and experiments in the absence of protein biosynthesis revealed that the nuclear import of viral ribonucleoprotein complexes (vRNPs) was already blocked in the absence of PEPD. These results led us to investigate which step during entry was affected. Receptor expression, IAV attachment, or IAV internalization was not dependent on the presence of PEPD. However, when looking at the distribution of incoming IAV particles in PEPD-knockdown cells, we found a localization pattern that differed from that in control cells: IAV mostly localized to the cell periphery, and consequently, viral particles displayed reduced colocalization with early and late endosome markers and fusion between viral and endosomal membranes was strongly reduced. Finally, experiments using a competitive inhibitor of PEPD catalytic activity suggested that the enzymatic function of the dipeptidase is required for its proviral effect on IAV entry. In sum, this study establishes PEPD as a novel entry factor required for early endosomal trafficking of IAV. Influenza A virus (IAV) continues to be a constant threat to public health. As IAV relies on its host cell for replication, the identification of host factors required by the virus is of importance. First, such studies often reveal novel functions of cellular factors and can extend our knowledge of cellular processes. Second, we can further our understanding of processes that are required for the entry of IAV into target cells. Third, the identification of host factors that contribute to IAV entry will increase the number of potential targets for the development of novel antiviral drugs that are of urgent need. Our study identifies prolidase (PEPD) to be a novel entry factor required by IAV for correct routing within the endosomal compartment following virus internalization. Thereby, we link PEPD, which has been shown to play a role during collagen recycling and growth factor signaling, to early events of viral infection.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Actidion, from microbial, ≥94% (TLC)
Sigma-Aldrich
Cycloheximid -Lösung, Ready-Made Solution, microbial, 100 mg/mL in DMSO, Suitable for cell culture
Sigma-Aldrich
Cycloheximid, ≥95% (HPLC)
Sigma-Aldrich
Actidion, Biotechnology Performance Certified
Millipore
Cycloheximid -Lösung, 0.1%, suitable for microbiology
Sigma-Aldrich
Prolidase aus Schweineniere, lyophilized powder, ≥100 units/mg protein
Sigma-Aldrich
2-Phenylindol, technical grade, 95%
Sigma-Aldrich
Z-Pro-OH, 99%
Supelco
Actidion, PESTANAL®, analytical standard