Merck
Cadmium resistance in extremely halophilic archaeon Haloferax strain BBK2.
Chemosphere (2014-07-23)
Deepthi Das, Bhakti B Salgaonkar, Kabilan Mani, Judith M Braganca
ZUSAMMENFASSUNG
Halophilic archaea are prevalent in highly saline habitats. Haloferax strain BBK2 is an orange pigmented, exopolysaccharide (EPS) producing extremely halophilic archaeon, isolated from solar salterns of Ribandar, Goa, India. It grew in varying pH (5-10) and NaCl concentration (10-30%). The isolate grew well in complex (NTYE) and minimal media (NGSM) in presence of heavy metal cadmium (Cd) up to 4.0 mM (805.28 mg L(-1)) concentration. The optimum growth in the presence and absence of Cd was seen at a pH range of 7-9 and salinity of 15-25%. The growth kinetics of the isolate in NTYE showed a specific growth rate (μmax) of 0.352 with generation time of 1.968 days. In presence of 1mM Cd, the μmax was 0.325 day(-1) and generation time was 2.132 days. In NGSM, the μmax decreased from 0.517 day(-1) (in control) to 0.265 day(-1) in 1mM Cd while, the doubling time increased from 1.34 days in control to 2.615 days in presence of 1 mM Cd. SDS PAGE of the whole cell protein extracts showed overexpressed proteins of 74.14 and 40 kDa. The scanning electron microscopy, energy dispersive X-ray spectroscopy (SEM-EDX) analysis of the intact cells and cells disrupted by dialysis revealed that Cd was bound onto the cells, which was further confirmed by AAS, FTIR and XRD analysis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cadmium, granular, 30-80 mesh, ≥99%
Sigma-Aldrich
Cadmium, powder, −100 mesh, 99.5% trace metals basis
Sigma-Aldrich
Cadmium, granular, ≥99%, 5-20 mesh
Sigma-Aldrich
Cadmium, shot, 3 mm, 99.999% trace metals basis
Cadmium, foil, 100x100mm, thickness 1.0mm, as rolled, 99.99%
Cadmium, foil, 10mm disks, thickness 1.0mm, as rolled, 100%
Cadmium, rod, 100mm, diameter 40mm, 99.9%
Cadmium, rod, 25mm, diameter 40mm, 99.9%
Cadmium, rod, 100mm, diameter 10.0mm, 99.999%
Cadmium, foil, 100x100mm, thickness 0.25mm, as rolled, 99.99+%
Cadmium, foil, 2m coil, thickness 0.5mm, as rolled, 100%
Cadmium, wire reel, 1m, diameter 1.2mm, as drawn, 99.99+%
Cadmium, rod, 200mm, diameter 10.0mm, 99.999%
Cadmium, rod, 50mm, diameter 10.0mm, 99.999%
Cadmium, foil, 25x25mm, thickness 0.5mm, as rolled, 99.999%
Cadmium, foil, 25x25mm, thickness 1.0mm, as rolled, 99.999%
Cadmium, foil, 50x50mm, thickness 0.25mm, as rolled, 99.9%
Cadmium, foil, 150x150mm, thickness 0.5mm, as rolled, 99.99%
Cadmium, foil, 30x50mm, thickness 0.5mm, as rolled, 99.999%
Cadmium, foil, 50x50mm, thickness 0.125mm, as rolled, 99.99+%
Cadmium, tube, 500mm, outside diameter 2.29mm, inside diameter 1.27mm, wall thickness 0.51mm, 99.95+%
Cadmium, foil, 100x300mm, thickness 0.125mm, as rolled, 99.95%
Cadmium, foil, 25x25mm, thickness 0.25mm, as rolled, 99.9%
Cadmium, foil, 50x50mm, thickness 0.25mm, as rolled, 99.99+%
Cadmium, foil, 50x50mm, thickness 2.0mm, as rolled, 99.99%
Cadmium, foil, 4mm disks, thickness 0.5mm, as rolled, 99.95%
Cadmium, foil, 8mm disks, thickness 0.005mm, 99.7+%
Cadmium, foil, not light tested, 50x50mm, thickness 0.005mm, 99.7+%
Cadmium, wire reel, 0.1m, diameter 1.2mm, as drawn, 99.99+%
Cadmium, foil, 100x100mm, thickness 0.125mm, as rolled, 99.99+%