Merck
  • The inverse agonist propranolol confers no corticosteroid-sparing activity in mild-to-moderate persistent asthma.
The inverse agonist propranolol confers no corticosteroid-sparing activity in mild-to-moderate persistent asthma.
Clinical science (London, England : 1979) (2014-06-19)
William J Anderson, Philip M Short, Peter A Williamson, Arvind Manoharan, Brian J Lipworth
ZUSAMMENFASSUNG
The murine asthma model shows that switching off airway β2 receptors with an inverse agonist may confer anti-inflammatory effects as well as corticosteroid-sparing activity. We have assessed for any corticosteroid-sparing effects of propranolol, an inverse agonist, added to low-dose inhaled corticosteroid (ICS) compared with higher dose ICS. A randomized double-blind placebo-controlled cross-over trial in mild-to-moderate persistent asthmatic patients was performed. After a run-in (2 weeks) on hydrofluoroalkane-beclometasone dipropionate (HFA-BDP) at 100 μg/day (HFA-BDP100), patients received randomized treatments (4 weeks) with propranolol at 80 mg/day plus HFA-BDP at 100 μg/day compared with placebo plus HFA-BDP at 400 μg/day (HFA-BDP400). Propranolol was up-titrated to 80 mg/day over the initial 2 weeks. Tiotropium was co-administered until 5 days before each histamine challenge (the primary outcome). Sixteen patients completed the study [mean age, 38 years; forced expiratory volume in 1 s (FEV1), 86.4%; histamine provocative concentration causing a 20% fall in FEV1 (PC20), 1.39 mg/ml; ICS dose, 406 μg/day]. Histamine PC20 was unchanged by adding propranolol to HFA-BDP100 compared with baseline (HFA-BDP100) {0.17 doubling dilution (dd) difference [95% confidence interval (CI): -0.58 to 0.92]}, but there was a significant improvement with HFA-BDP400 compared with both baseline [1.05 dd (95% CI: 0.43-1.66); P=0.02], and propranolol+HFA-BDP100 [0.88 dd (95% CI: 0.45-1.30); P=0.006]. Significant improvements were also observed with HFA-BDP400 for exhaled nitric oxide, blood eosinophils, serum eosinophilic cationic protein and asthma quality-of-life questionnaire symptoms compared with propranolol+HFA-BDP100. Salbutamol recovery post-challenge was partially blunted by propranolol (median prolongation 5 min; P=0.002). Domiciliary evening FEV1 also fell with propranolol+HFA-BDP100 [mean reduction from baseline 0.22 litres (95% CI: 0.10-0.34); P=0.012], whereas Asthma Control Questionnaire remained unchanged. In conclusion, the inverse agonist propranolol produced no improvements when given with low-dose ICS, whereas further significant improvements in airway hyper-responsiveness and inflammation were demonstrated with higher dose ICS. Thus, propranolol does not confer corticosteroid-sparing activity in persistent asthma.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Hydrocortison, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortison -Lösung, 50 μM, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrocortison, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrocortison, ≥98% (HPLC)
Sigma-Aldrich
Kreatinin, anhydrous, ≥98%
Sigma-Aldrich
(±)-Propranolol -hydrochlorid, ≥99% (TLC), powder
Supelco
Cortisol -Lösung, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Hydrocortison, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Hydrocortison, United States Pharmacopeia (USP) Reference Standard
Supelco
Propranolol-Hydrochlorid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Beclomethason, ≥99%
USP
Propranolol -hydrochlorid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hydrocortison, meets USP testing specifications
Supelco
Propranolol -hydrochlorid, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Propranolol -hydrochlorid, European Pharmacopoeia (EP) Reference Standard
Hydrocortison für die Peakidentifizierung, European Pharmacopoeia (EP) Reference Standard
Supelco
(±)-Propranolol -hydrochlorid, analytical standard
Hydrocortison, European Pharmacopoeia (EP) Reference Standard