Merck
Transformation and removal pathways of four common PPCP/EDCs in soil.
Environmental pollution (Barking, Essex : 1987) (2014-07-06)
L K Dodgen, J Li, X Wu, Z Lu, J J Gan
ZUSAMMENFASSUNG
Pharmaceutical and personal care products (PPCPs) and endocrine disrupting chemicals (EDCs) enter the soil environment via irrigation with treated wastewater, groundwater recharge, and land application of biosolids. The transformation and fate of PPCP/EDCs in soil affects their potential for plant uptake and groundwater pollution. This study examined four PPCP/EDCs (bisphenol A, diclofenac, naproxen, and 4-nonylphenol) in soil by using (14)C-labeling and analyzing mineralization, extractable residue, bound residue, and formation of transformation products. At the end of 112 d of incubation, the majority of (14)C-naproxen and (14)C-diclofenac was mineralized to (14)CO2, while a majority of (14)C-bisphenol A and (14)Cnonylphenol was converted to bound residue. After 112 d, the estimated half-lives of the parent compounds were only 1.4-5.4 d. However a variety of transformation products were found and several for bisphenol A and diclofenac were identified, suggesting the need to consider degradation intermediates in soils impacted by PPCP/EDCs.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
4,4′-Isopropylidendiphenol, ≥99%
Sigma-Aldrich
Diclofenac Natriumsalz
Supelco
Diclofenac Natriumsalz, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Naproxen, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
4,4′-Isopropylidendiphenol, 97%
USP
Diclofenacnatrium, United States Pharmacopeia (USP) Reference Standard
Supelco
4,4′-Isopropylidendiphenol, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
USP
Naproxen, United States Pharmacopeia (USP) Reference Standard
Supelco
Diclofenac Natriumsalz, analytical standard
Sigma-Aldrich
Naproxen, meets USP testing specifications
Supelco
4-Nonylphenol, PESTANAL®, analytical standard
Diclofenac für die Systemeignung, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
(S)-(+)-6-Methoxy-α-methyl-2-naphthalinessigsäure, 98%
Diclofenac-Natrium, European Pharmacopoeia (EP) Reference Standard
Supelco
Naproxen, VETRANAL®, analytical standard
Supelco
Naproxen -Lösung, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Naproxen, European Pharmacopoeia (EP) Reference Standard
Diclofenac-Kalium, European Pharmacopoeia (EP) Reference Standard