Merck
  • Accumulation of cadmium in and its effect on the midgut gland of terrestrial snail Helix pomatia L. from urban areas in Poland.
Accumulation of cadmium in and its effect on the midgut gland of terrestrial snail Helix pomatia L. from urban areas in Poland.
Bulletin of environmental contamination and toxicology (2014-08-13)
Tadeusz Włostowski, Paweł Kozłowski, Barbara Laszkiewicz-Tiszczenko, Ewa Oleńska, Olgierd Aleksandrowicz
ZUSAMMENFASSUNG
The objectives of this study were (1) to determine cadmium (Cd) accumulation in the midgut gland of a land snail Helix pomatia L. inhabiting residential areas of the 14 largest cities in Poland, and (2) to examine whether the accumulated Cd exerted any toxic effects. The average accumulation of Cd in the midgut gland of snails, weighing 16-18 g, ranged from 7.00 to 87.3 µg/g dry weight (0.06-0.77 µmol/g) and differed significantly among animals from the various urban areas. This difference in Cd accumulation was not related to city population, but was associated with the topsoil Cd (R(2) = 0.868, p < 0.0001). The tissue Cd was not found to produce toxicity (histopathology, programmed cell death, lipofuscin formation or lipid peroxidation), probably due to the induction of sufficiently high quantities of metallothionein and glutathione, well-known protective molecules.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Glutathion reduziert, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathion reduziert, ≥98.0%
Supelco
Glutathion, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutathion reduziert, BioXtra, ≥98.0%
Sigma-Aldrich
Cadmium, granular, 30-80 mesh, ≥99%
Sigma-Aldrich
Cadmium, powder, −100 mesh, 99.5% trace metals basis
Sigma-Aldrich
Cadmium, granular, ≥99%, 5-20 mesh
Glutathion, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Cadmium, shot, 3 mm, 99.999% trace metals basis
Cadmium, foil, 100x100mm, thickness 1.0mm, as rolled, 99.99%
Cadmium, foil, 10mm disks, thickness 1.0mm, as rolled, 100%
Cadmium, rod, 100mm, diameter 40mm, 99.9%
Cadmium, rod, 25mm, diameter 40mm, 99.9%
Cadmium, rod, 100mm, diameter 10.0mm, 99.999%
Cadmium, foil, 100x100mm, thickness 0.25mm, as rolled, 99.99+%
Cadmium, foil, 2m coil, thickness 0.5mm, as rolled, 100%
Cadmium, wire reel, 1m, diameter 1.2mm, as drawn, 99.99+%
Cadmium, rod, 200mm, diameter 10.0mm, 99.999%
Cadmium, rod, 50mm, diameter 10.0mm, 99.999%
Cadmium, foil, 25x25mm, thickness 0.5mm, as rolled, 99.999%
Cadmium, foil, 25x25mm, thickness 1.0mm, as rolled, 99.999%
Cadmium, foil, 50x50mm, thickness 0.25mm, as rolled, 99.9%
Cadmium, foil, 150x150mm, thickness 0.5mm, as rolled, 99.99%
Cadmium, foil, 30x50mm, thickness 0.5mm, as rolled, 99.999%
Cadmium, foil, 50x50mm, thickness 0.125mm, as rolled, 99.99+%
Cadmium, tube, 500mm, outside diameter 2.29mm, inside diameter 1.27mm, wall thickness 0.51mm, 99.95+%
Cadmium, foil, 100x300mm, thickness 0.125mm, as rolled, 99.95%
Cadmium, foil, 25x25mm, thickness 0.25mm, as rolled, 99.9%
Cadmium, foil, 50x50mm, thickness 0.25mm, as rolled, 99.99+%
Cadmium, foil, 50x50mm, thickness 2.0mm, as rolled, 99.99%