Merck
Chiral speciation of selenoamino acids in biological samples.
Journal of chromatography. A (2014-08-19)
Beibei Chen, Man He, Cheng Zhong, Bin Hu
ZUSAMMENFASSUNG
In this paper, the "state of the art" of chiral speciation of selenoamino acids (SeAAs) in biological samples is critically reviewed. The significance and the features of such studies are highlighted. A special focus lies on chiral speciation of SeAAs by hyphenation techniques in which a chiral separation method (such as gas chromatography (GC), high performance liquid chromatography (HPLC) and capillary electrophoresis (CE)) is on-line coupled with an elemental specific detector, especially inductively coupled plasma mass spectrometry (ICP-MS). The advances in the development and application of hyphenation techniques in chiral speciation of SeAAs in biological samples are summarized and a perspective for future developments including sophisticated and innovative applications is discussed. Overall, HPLC-ICP-MS is more applicable than GC/CE-ICP-MS for chiral speciation of SeAAs. In the future, more novel chiral HPLC methods with high enantio-resolution, low cost and robustness, and their more applications in real biological samples analysis are expected.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methansulfonsäure, ≥99.0%
Sigma-Aldrich
Selen, powder, −100 mesh, 99.99% trace metals basis
Supelco
Astec® CHIROBIOTIC® T Chirale HPLC-Säule, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Sigma-Aldrich
Selen, powder, −100 mesh, ≥99.5% trace metals basis
Supelco
Astec® CYCLOBOND I 2000 Chirale HPLC-Säule, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
Methansulfonsäure, suitable for HPLC, LiChropur, ≥99.5% (T)
Supelco
Methansulfonsäurekonzentrat, 0.1 M CH3SO3H in water (0.1N), eluent concentrate for IC
Sigma-Aldrich
Methansulfonsäure -Lösung, 70 wt. % in H2O
Sigma-Aldrich
Selen, pellets, <5 mm particle size, ≥99.999% trace metals basis
Sigma-Aldrich
Selen, pellets, <5 mm, ≥99.99% trace metals basis
Supelco
Astec® CHIROBIOTIC® T Chirale HPLC-Säule, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
Astec® CHIROBIOTIC® T Chirale HPLC-Säule, 5 μm particle size, L × I.D. 15 cm × 2.1 mm
Supelco
Astec® CHIROBIOTIC® T Chirale HPLC-Säule, 5 μm particle size, L × I.D. 25 cm × 2.1 mm
Supelco
Astec® CHIROBIOTIC® T Chirale HPLC-Säule, 5 μm particle size, L × I.D. 10 cm × 2.1 mm
Sigma-Aldrich
Methansulfonsäure -Lösung, 4 M (with 0.2% (w/v) tryptamine)
Selen, foil, 25x25mm, thickness 3mm, 99.95%
Supelco
Astec® CHIROBIOTIC® T Chirale HPLC-Säule, 5 μm particle size, L × I.D. 5 cm × 4.6 mm
Supelco
Astec® CYCLOBOND I 2000 Chirale HPLC-Säule, 5 μm particle size, L × I.D. 10 cm × 2.1 mm
Supelco
Astec® CYCLOBOND I 2000 Chirale HPLC-Säule, 5 μm particle size, L × I.D. 15 cm × 2.1 mm
Supelco
Astec® CYCLOBOND I 2000 Chirale HPLC-Säule, 5 μm particle size, L × I.D. 25 cm × 10 mm
Selen, pellets, < 5mm, ≥99.999%
Supelco
Astec® CHIROBIOTIC® T Chirale HPLC-Säule, 5 μm particle size, L × I.D. 10 cm × 4.6 mm