Merck
  • Organotropism of persistent organic pollutants and heavy metals in the Greenland shark Somniosus microcephalus in NE Greenland.
Organotropism of persistent organic pollutants and heavy metals in the Greenland shark Somniosus microcephalus in NE Greenland.
Marine pollution bulletin (2014-08-03)
Simonetta Corsolini, Stefania Ancora, Nicola Bianchi, Giacomo Mariotti, Claudio Leonzio, Jørgen S Christiansen
ZUSAMMENFASSUNG
The Greenland shark Somniosus microcephalus is an opportunistic feeder, a top predator, and a very long-lived species. The brain, liver, red and white muscle, gonads, fat, skin, pancreas, and spleen of Greenland sharks from NE Greenland fjords were analysed for PCBs, PCDDs/DFs, PBDEs; DDT isomers; HCH isomers; dieldrin; endrin; HCB; Cd, Hg, Pb, and Se. PCBs (2.01-103 ng/g wet wt) and PBDEs (7.9-3050 pg/g wet wt) were detected in most of the samples. PCDDs/DFs showed high values when detected. DDTs, HCB and HCHs were only detected in some tissues. The ΣTEQ was 5.76 pg/g in muscle. Cadmium mainly accumulated in the pancreas and liver (19.6 and 10.7 mg/kg dry wt, respectively); mercury in red muscle (4.10-6.91 mg/kg dry wt); selenium in the pancreas (3.57 mg/kg dry wt) and spleen (1.95 mg/kg dry wt); lead in the skin (0.358 mg/kgd ry wt). The selenium-mercury ratio in the liver was also evaluated.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Selen, powder, −100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Selen, powder, −100 mesh, ≥99.5% trace metals basis
Sigma-Aldrich
Quecksilber, ACS reagent, 99.9995% trace metals basis
Sigma-Aldrich
Cadmium, granular, 30-80 mesh, ≥99%
Sigma-Aldrich
Cadmium, powder, −100 mesh, 99.5% trace metals basis
Sigma-Aldrich
Selen, pellets, <5 mm particle size, ≥99.999% trace metals basis
Sigma-Aldrich
Cadmium, granular, ≥99%, 5-20 mesh
Sigma-Aldrich
Selen, pellets, <5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Cadmium, shot, 3 mm, 99.999% trace metals basis
Cadmium, foil, 100x100mm, thickness 1.0mm, as rolled, 99.99%
Selen, foil, 25x25mm, thickness 3mm, 99.95%
Cadmium, foil, 10mm disks, thickness 1.0mm, as rolled, 100%
Cadmium, rod, 100mm, diameter 40mm, 99.9%
Cadmium, rod, 25mm, diameter 40mm, 99.9%
Cadmium, rod, 100mm, diameter 10.0mm, 99.999%
Cadmium, foil, 100x100mm, thickness 0.25mm, as rolled, 99.99+%
Selen, pellets, < 5mm, ≥99.999%
Cadmium, foil, 2m coil, thickness 0.5mm, as rolled, 100%
Cadmium, wire reel, 1m, diameter 1.2mm, as drawn, 99.99+%
Cadmium, rod, 200mm, diameter 10.0mm, 99.999%
Cadmium, rod, 50mm, diameter 10.0mm, 99.999%
Cadmium, foil, 25x25mm, thickness 0.5mm, as rolled, 99.999%
Cadmium, foil, 25x25mm, thickness 1.0mm, as rolled, 99.999%
Cadmium, foil, 50x50mm, thickness 0.25mm, as rolled, 99.9%
Cadmium, foil, 150x150mm, thickness 0.5mm, as rolled, 99.99%
Cadmium, foil, 30x50mm, thickness 0.5mm, as rolled, 99.999%
Cadmium, foil, 50x50mm, thickness 0.125mm, as rolled, 99.99+%
Cadmium, tube, 500mm, outside diameter 2.29mm, inside diameter 1.27mm, wall thickness 0.51mm, 99.95+%
Cadmium, foil, 100x300mm, thickness 0.125mm, as rolled, 99.95%
Cadmium, foil, 25x25mm, thickness 0.25mm, as rolled, 99.9%