Merck
  • Bacterial maximum non-inhibitory and minimum inhibitory concentrations of different water activity depressing solutes.
Bacterial maximum non-inhibitory and minimum inhibitory concentrations of different water activity depressing solutes.
International journal of food microbiology (2014-08-05)
G Cebrián, C Arroyo, P Mañas, S Condón
ZUSAMMENFASSUNG
The NaCl MNICs (maximum non-inhibitory concentrations) and MICs (minimum inhibitory concentrations) for growth of various strains of six bacterial species were determined and then compared with those obtained for seven other solutes. The influence of prior growth conditions on the MNICs and MICs was also evaluated. No significant changes on the MNICs and MICs were found among the strains studied within each species. Among all factors investigated, only growth phase -for Gram-negatives- and growth at high NaCl concentrations led to a change in the NaCl MNICs. Species could be classified depending on its NaCl MNICs and MICs (in decreasing order) as follows: Staphylococcus aureus, Listeria monocytogenes, Cronobacter sakazakii, Enterococcus faecium, Escherichia coli and Salmonella Typhimurium. Similar results were obtained for KCl, LiCl, and sodium acetate, but not for the remaining solutes investigated (sucrose, glycerol, MgCl2 and CaCl2). Results obtained indicate that, in general, Gram-negatives showed lower MNICs and MICs than Gram-positives for all the solutes, S. aureus being the most solute tolerant microorganism. When compared on a molar basis, glycerol showed the highest MNICs and MICs for all the microorganisms -except for S. aureus- and LiCl the lowest ones. NaCl MNICs and MICs were not significantly different from those of KCl when compared on a molar basis. Therefore, the inhibitory action of NaCl could not be linked to the specific action of Na(+). Results also showed that the Na(+) tolerance of some species was Cl(-) dependent whereas for others it was not, and that factors others than aw-decrease contribute to the inhibitory action of LiCl, CaCl2 and MgCl2.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Wasser, suitable for HPLC
Sigma-Aldrich
Glycerin, ACS reagent, ≥99.5%
Sigma-Aldrich
Wasser, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Glycerin, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerin, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Saccharose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Wasser, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Lithiumchlorid, ACS reagent, ≥99%
Sigma-Aldrich
Calciumchlorid Dihydrat, ACS reagent, ≥99%
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Magnesiumchlorid -Lösung, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Calciumchlorid Dihydrat, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Wasser, Deionized
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Magnesiumchlorid, ≥98%
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Saccharose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Calciumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Glycerin, ≥99.5%
Supelco
Wasser, suitable for ion chromatography
Sigma-Aldrich
Calciumchlorid Dihydrat, Molecular Biology, ≥99.0%
Supelco
Saccharose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Lithiumchlorid, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
USP
Saccharose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Calciumchlorid, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calciumchlorid Dihydrat, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%