Merck
  • Elucidation of kinetic mechanisms of human translesion DNA polymerase κ using tryptophan mutants.
Elucidation of kinetic mechanisms of human translesion DNA polymerase κ using tryptophan mutants.
The FEBS journal (2014-07-30)
Linlin Zhao, Matthew G Pence, Robert L Eoff, Shuai Yuan, Catinca A Fercu, F Peter Guengerich
ZUSAMMENFASSUNG
To investigate the conformational dynamics of human DNA polymerase κ (hpol κ), we generated two mutants, Y50W (N-clasp region) and Y408W (linker between the thumb and little finger domains), using a Trp-null mutant (W214Y/W392H) of the hpol κ catalytic core enzyme. These mutants retained catalytic activity and similar patterns of selectivity for bypassing the DNA adduct 7,8-dihydro-8-oxo-2'-deoxyguanosine, as indicated by the results of steady-state and pre-steady-state kinetic experiments. Stopped-flow kinetic assays with hpol κ Y50W and T408W revealed a decrease in Trp fluorescence with the template G:dCTP pair but not for any mispairs. This decrease in fluorescence was not rate-limiting and is considered to be related to a conformational change necessary for correct nucleotidyl transfer. When a free 3'-hydroxyl was present on the primer, the Trp fluorescence returned to the baseline level at a rate similar to the observed kcat , suggesting that this change occurs during or after nucleotidyl transfer. However, polymerization rates (kpol ) of extended-product formation were fast, indicating that the slow fluorescence step follows phosphodiester bond formation and is rate-limiting. Pyrophosphate formation and release were fast and are likely to precede the slower relaxation step. The available kinetic data were used to fit a simplified minimal model. The extracted rate constants confirmed that the conformational change after phosphodiester bond formation was rate-limiting for hpol κ catalysis with the template G:dCTP pair.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Taq-DNA-Polymerase aus Thermus aquaticus, with 10× PCR reaction buffer containing MgCl2
Sigma-Aldrich
Taq-DNA-Polymerase aus Thermus aquaticus, with 10× PCR reaction buffer without MgCl2
Sigma-Aldrich
DNA-Polymerase I, Klenow-Fragment aus E. coli, buffered aqueous glycerol solution
Sigma-Aldrich
DNA-Polymerase I aus E. coli lysogen für NM 964, buffered aqueous glycerol solution