Merck
  • The cross-priming capacity and direct presentation potential of an autoantigen are separable and inversely related properties.
The cross-priming capacity and direct presentation potential of an autoantigen are separable and inversely related properties.
Journal of immunology (Baltimore, Md. : 1950) (2014-08-29)
Jinguo Wang, Roopa Hebbandi Nanjundappa, Afshin Shameli, Xavier Clemente-Casares, Jun Yamanouchi, John F Elliott, Robyn Slattery, Pau Serra, Pere Santamaria
ZUSAMMENFASSUNG
We investigated whether a prevalent epitope of the β-cell-specific autoantigen islet-specific glucose-6-phosphatase catalytic subunit-related protein (IGRP206-214) reaches regional Ag-presentation pathways via unprocessed polypeptide chains, as free IGRP206-214 peptide or via preformed IGRP206-214/K(d) complexes. This was accomplished by expressing bacterial artificial chromosome transgenes encoding wild-type (stable) or ubiquitinated (unstable) forms of IGRP in IGRP-deficient NOD mice carrying MHC class I-deficient β-cells, dendritic cells, or B cells. We investigated the ability of the pancreatic lymph nodes of these mice to prime naive IGRP206-214-reactive CD8(+) T cells in vivo, either in response to spontaneous Ag shedding, or to synchronized forms of β-cell necrosis or apoptosis. When IGRP was made unstable by targeting it for proteasomal degradation within β-cells, the cross-priming, autoimmune-initiating potential of this autoantigen (designated autoantigenicity) was impaired. Yet at the same time, the direct presentation, CTL-targeting potential of IGRP (designated pathogenicity) was enhanced. The appearance of IGRP206-214 in regional Ag-presentation pathways was dissociated from transfer of IGRP206-214 or IGRP206-214/K(d) from β cells to dendritic cells. These results indicate that autoantigenicity and pathogenicity are separable and inversely related properties and suggest that pathogenic autoantigens, capable of efficiently priming CTLs while marking target cells for CTL-induced killing, may have a critical balance of these two properties.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Interleukin-2 human, IL-2, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
5(6)-Carboxyfluoreszeindiacetat-N-succinimidylester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Interleukin-2 human, IL-2, recombinant, expressed in HEK 293 cells, suitable for cell culture, endotoxin tested
Sigma-Aldrich
Interleukin-2 human, recombinant, expressed in Pichia pastoris, suitable for cell culture
Sigma-Aldrich
Humanes Interleukin-2 human, Animal-component free, recombinant, expressed in E. coli, suitable for cell culture
Sigma-Aldrich
Mouse IFN-γ ELISA Kit, for serum, plasma, cell culture supernatant
Sigma-Aldrich
Interleukin-2, recombinant, expressed in E. coli, ~10000 U/mL
Sigma-Aldrich
5-Carboxy-Fluoresceindiacetat N-Succinimidylester, suitable for fluorescence, ≥95.0% (HPLC)
Sigma-Aldrich
Bovine IFNG / Interferon Gamma ELISA Kit, for serum, plasma and cell culture supernatants
Sigma-Aldrich
Mouse IFN-γ ELISA Kit, for cell and tissue lysates
Sigma-Aldrich
Rat IFN-γ ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Cotton Rat Ifng / Interferon Gamma ELISA Kit