- Niacin for reduction of cardiovascular risk.
Niacin for reduction of cardiovascular risk.
The New England journal of medicine (2014-11-13)
Inge A M van den Oever, Michael T Nurmohamed, Willem F Lems
PMID25390752
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Nicotinsäure, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Nicotinsäure, European Pharmacopoeia (EP) Reference Standard