- Comment on "Oxytocin-mediated GABA inhibition during delivery attenuates autism pathogenesis in rodent offspring".
Comment on "Oxytocin-mediated GABA inhibition during delivery attenuates autism pathogenesis in rodent offspring".
Science (New York, N.Y.) (2014-10-11)
Victorio Bambini-Junior, Gustavo Della Flora Nunes, Tomasz Schneider, Carmem Gottfried
PMID25301610
ZUSAMMENFASSUNG
Tyzio et al. (Reports, 7 February 2014, p. 675) reported that bumetanide restored the impaired oxytocin-mediated γ-aminobutyric acid (GABA) excitatory-inhibitory shift during delivery in animal models of autism, ameliorating some autistic-like characteristics in the offspring. However, standard practices in the study of these models, such as the use of sex-dimorphic or males-only analyses and implementation of tests measuring social behavior, are lacking to definitely associate their findings to autism.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Oxytocin, European Pharmacopoeia (EP) Reference Standard
Vigabatrin Unreinheit D, European Pharmacopoeia (EP) Reference Standard