- The BTB/POZ domain: a new protein-protein interaction motif common to DNA- and actin-binding proteins.
The BTB/POZ domain: a new protein-protein interaction motif common to DNA- and actin-binding proteins.
Cell growth & differentiation : the molecular biology journal of the American Association for Cancer Research (1995-09-01)
O Albagli, P Dhordain, C Deweindt, G Lecocq, D Leprince
PMID8519696
ZUSAMMENFASSUNG
The BTB/POZ domain defines a newly characterized protein-protein interaction interface. It is highly conserved throughout metazoan evolution and generally found at the NH2 terminus of either actin-binding or, more commonly, nuclear DNA-binding proteins. By mediating protein binding in large aggregates, the BTB/POZ domain serves to organize higher order macromolecular complexes involved in ring canal formation or chromatin folding.