Merck
  • Efficient Reduction of Ethyl 2-Oxo-4-phenylbutyrate at 620 g? L- 1 by a Bacterial Reductase with Broad Substrate Spectrum.
Efficient Reduction of Ethyl 2-Oxo-4-phenylbutyrate at 620 g? L- 1 by a Bacterial Reductase with Broad Substrate Spectrum.
Advanced Synthesis & Catalysis (2011)
Ni Y, et al.