Merck
  • Knockdown of radixin by RNA interference suppresses the growth of human pancreatic cancer cells in vitro and in vivo.
Knockdown of radixin by RNA interference suppresses the growth of human pancreatic cancer cells in vitro and in vivo.
Asian Pacific journal of cancer prevention : APJCP (2012-05-29)
Shu-Dong Chen, Mao-Min Song, Zhi-Qiang Zhong, Na Li, Pi-Lin Wang, Shi Cheng, Ri-Xing Bai, Hui-Sheng Yuan
ZUSAMMENFASSUNG
Radixin, encoded by a gene on chromosome 11, plays important roles in cell motility, invasion and tumor progression. However, its function in pancreatic cancer remains elusive. In this study, radixin gene expression was suppressed with a lentivirus-mediated short-hairpin RNA (shRNA) method. We found that radixin shRNA caused down-regulation of radixin in PANC-1 cells, associated with inhibition of pancreatic cancer cell proliferation, survival, adhesion and invasive potential in vitro. When radixin-silenced cells were implanted in nude mice, tumor growth and microvessel density were significantly inhibited as compared to blank control cells or nonsense shRNA control cells. Thrombospondin-1 (TSP-1) and E-cadherin were up-regulated in radixin-silenced PANC-1 cells. Our results suggest that radixin might play a critical role in pancreatic cancer progression, possibly through involvement of down-regulation of TSP-1 and E-cadherin expression.