Merck
  • Antisense oligonucleotides from the stage-specific myeloid zinc finger gene MZF-1 inhibit granulopoiesis in vitro.
Antisense oligonucleotides from the stage-specific myeloid zinc finger gene MZF-1 inhibit granulopoiesis in vitro.
The Journal of experimental medicine (1991-11-01)
L Bavisotto, K Kaushansky, N Lin, R Hromas
ZUSAMMENFASSUNG
Zinc finger proteins are transcriptional regulators of other genes, often controlling developmental cascades of gene expression. A recently cloned zinc finger gene, MZF-1, was found to be preferentially expressed in myeloid cells. Using complementary radiolabeled MZF-1 RNA hybridized to human bone marrow smears in situ, it was discovered that the expression of MZF-1 is essentially limited to the myelocyte and metamyelocyte stages of granulopoiesis. Antisense but not sense oligonucleotides from MZF-1 significantly inhibited granulocyte colony-stimulating factor-driven granulocyte colony formation in vitro.