Merck
  • Generation of transparent conductive electrodes by laser consolidation of LIFT printed ITO nanoparticle layers.
Generation of transparent conductive electrodes by laser consolidation of LIFT printed ITO nanoparticle layers.
Applied Physics. A, Materials Science & Processing (2013)
Baum M, et al.