Merck
  • Methylmalonic semialdehyde dehydrogenase deficiency: psychomotor delay and methylmalonic aciduria without metabolic decompensation.
Methylmalonic semialdehyde dehydrogenase deficiency: psychomotor delay and methylmalonic aciduria without metabolic decompensation.
Molecular genetics and metabolism (1998-10-27)
C R Roe, E Struys, R M Kok, D S Roe, R A Harris, C Jakobs
ZUSAMMENFASSUNG
A patient presenting with developmental delay but no episodes of metabolic acidosis was found to excrete significant amounts of methylmalonate (MMA) without any associated increased excretion of malonate, ethylmalonate, 3-hydroxypropionate, or beta-alanine. In contrast to patients with methylmalonic aciduria due to deficient mutase or impaired cobalamin metabolism, there was no increase of propionylcarnitine in blood or urine. The activity of methylmalonyl-CoA mutase and the pathway for cobalamin metabolism were also intact. The quantitative levels of the various labeled enantiomers of 3-hydroxyisobutyric (3-HIBA), 3-aminoisobutyric (3-AIBA), MMA, and propionylcarnitine were compared following separate intravenous infusions of equimolar doses of [2H8]-valine or [2H4]thymine in this patient and another with methylmalonyl-CoA mutase deficiency. Levels of labeled S- and R-3-HIBA and S- and R-3-AIBA indicated an isolated defect in methylmalonic semialdehyde dehydrogenase in this patient. This condition can be recognized by plasma MMA levels of approximately 8.5 microM (cf. 400 microM in mutase deficiency), urine MMA of 20-55 micromol/kg/24 h (cf. 1150 micromol/kg/24 h), no increase in propionylcarnitine following an oral carnitine load, and increased excretion of S-3-AIBA-nearly 10 times that observed in mutase deficiency. The ratio of R-AIBA to S-AIBA of <1 also reflects this disorder.