Merck
  • Comparative HPLC methods for β-blockers separation using different types of chiral stationary phases in normal phase and polar organic phase elution modes. Analysis of propranolol enantiomers in natural waters.
Comparative HPLC methods for β-blockers separation using different types of chiral stationary phases in normal phase and polar organic phase elution modes. Analysis of propranolol enantiomers in natural waters.
Journal of pharmaceutical and biomedical analysis (2012-01-24)
Sonia Morante-Zarcero, Isabel Sierra
ZUSAMMENFASSUNG
The enantioselectivities of β-blockers (propranolol, metoprolol, atenolol and pindolol) on four different types of chiral stationary phases (CSPs): Chiralpak AD-H, Lux Cellulose-1, Chirobiotic T and Sumichiral OA-4900 were compared using polar organic (PO) elution mode and normal phase (NP) elution mode. Method optimizations were demonstrated by modifying parameters such as organic modifier composition (ethanol, 2-propanol and acetonitrile) and basic mobile phase additives (triethylamine, diethylamine, ethanolamine, and buthylamine). In normal phase elution mode with Lux Cellulose-1, the four pairs of enantiomers can be separated in the same run in gradient elution mode. Additionally, a simple chiral HPLC-DAD method using a newly commercialized polysaccharide-based CSP by Phenomenex (Lux Cellulose-1) in NP elution mode for enantioselective determination of propranolol in water samples by highly selective molecularly imprinted polymers extraction was validated. The optimized conditions were a mobile phase composed by n-hexane/ethanol/DEA (70/30/0.3, v/v/v) at a flow rate of 1.0 mL min(-1) and 25 °C. The method is selective, precise and accurate and was found to be linear in the range of 0.125-50 μg mL(-1) (R(2)>0.999) with a method detection limit (MLD) of 0.4 μg mL(-1) for both enantiomers. Recoveries achieved with both enantiomers ranged from 97 to 109%.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Astec® CHIROBIOTIC® T Chirale HPLC-Säule, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
Astec® CHIROBIOTIC® V Chirale HPLC-Säule, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
Astec® CHIROBIOTIC® T Chirale HPLC-Säule, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
Astec® CHIROBIOTIC® T Chirale HPLC-Säule, 5 μm particle size, L × I.D. 15 cm × 2.1 mm
Supelco
Astec® CHIROBIOTIC® T Chirale HPLC-Säule, 5 μm particle size, L × I.D. 25 cm × 2.1 mm
Supelco
Astec® CHIROBIOTIC® T Chirale HPLC-Säule, 5 μm particle size, L × I.D. 10 cm × 2.1 mm
Supelco
Astec® CHIROBIOTIC® V Chirale HPLC-Säule, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
Astec® CHIROBIOTIC® V Chirale HPLC-Säule, 5 μm particle size, L × I.D. 10 cm × 4.6 mm
Supelco
Astec® CHIROBIOTIC® V Chirale HPLC-Säule, 5 μm particle size, L × I.D. 15 cm × 2.1 mm
Supelco
Astec® CHIROBIOTIC® T Chirale HPLC-Säule, 5 μm particle size, L × I.D. 5 cm × 4.6 mm
Supelco
Astec® CHIROBIOTIC® T Chirale HPLC-Säule, 5 μm particle size, L × I.D. 25 cm × 10 mm
Supelco
Astec® CHIROBIOTIC® V Chirale HPLC-Säule, 5 μm particle size, L × I.D. 25 cm × 2.1 mm
Supelco
Astec® CHIROBIOTIC® V Chirale HPLC-Säule, 5 μm particle size, L × I.D. 10 cm × 2.1 mm
Supelco
Astec® CHIROBIOTIC® T Chirale HPLC-Säule, 5 μm particle size, L × I.D. 10 cm × 4.6 mm