Merck
  • A new X-linked syndrome with agenesis of the corpus callosum, mental retardation, coloboma, micrognathia, and a mutation in the Alpha 4 gene at Xq13.
A new X-linked syndrome with agenesis of the corpus callosum, mental retardation, coloboma, micrognathia, and a mutation in the Alpha 4 gene at Xq13.
American journal of medical genetics. Part A (2003-10-14)
John M Graham, Patricia Wheeler, Darci Tackels-Horne, Angela E Lin, Bryan D Hall, Melanie May, Kieran M Short, Charles E Schwartz, Timothy C Cox
ZUSAMMENFASSUNG
We describe two brothers with a unique pattern of malformations that includes coloboma (iris, optic nerve), high forehead, severe retrognathia, mental retardation, and agenesis of the corpus callosum (ACC). Both boys have low-set cupped ears with sensorineural hearing loss, normal phallus, pectus excavatum, scoliosis, and short stature. One brother had choanal atresia and cardiac defects consisting of ventricular septal defect (VSD) and patent ductus arteriosus (PDA) which resolved spontaneously. Differential diagnosis between a number of clinical entities was considered, however, because ACC and the distinctive facial features were reminiscent of FG syndrome, DNA was analyzed for markers linked to the FGS1 locus at Xq13-q21. Notably, the brothers were concordant for markers spanning this presumed FG region, and in both we have identified adjacent alterations (-57delT and T-55A) in the Alpha 4 gene located within this interval. Alpha 4 is a regulatory subunit of the major cellular phosphatase, PP2A, that has recently been shown to interact with MID1, the product of the gene mutated in X-linked Opitz GBBB syndrome. The double nucleotide change identified in this family was not observed in 410 control chromosomes, suggesting that it may be a pathogenetic change. Altered expression of Alpha 4, through either a change in translational efficiency, mRNA stability or splicing, could explain the clinical phenotype in these boys and the phenotypic overlap with Opitz GBBB syndrome.