Merck
Genetic analysis of the first 4 patients with beta-ureidopropionase deficiency.
Nucleosides, nucleotides & nucleic acids (2006-10-27)
A B P van Kuilenburg, R Meinsma, B Assman, G F Hoffman, T Voit, A Ribes, I Lorente, R Busch, E Mayatepek, N G G M Abeling, R A Wevers, F Rutsch, A H van Gennip
ZUSAMMENFASSUNG
beta-Ureidopropionase is the third enzyme of the pyrimidine degradation pathway and it catalyses the irreversible hydrolysis of N-carbamyl-ss-aminoisobutyric acid or N-carbamyl-ss-alanine to beta-aminoisobutyric acid or ss-alanine, ammonia, and CO2. Analysis of the beta-ureidopropionase gene (UPB1) of the first 4 patients presenting with a complete enzyme deficiency, revealed the presence of 2 splice-site mutations (IVS1-2A>G and IVS8-1G>A) and one missense mutation (A85E). RT-PCR analysis of the complete beta-ureidopropionase cDNA suggested that both splice-site mutations lead to a variety of alternative splice variants, with deletions of a single or several exons. The alanine at position 85 was not conserved in other eukaryotic beta-ureidopropionase protein sequences.