Merck
Microfluidic perfusion culture system for multilayer artery tissue models.
Biomicrofluidics (2015-01-02)
Yuka Yamagishi, Taisuke Masuda, Michiya Matsusaki, Mitsuru Akashi, Utako Yokoyama, Fumihito Arai
ZUSAMMENFASSUNG
We described an assembly technique and perfusion culture system for constructing artery tissue models. This technique differed from previous studies in that it does not require a solid biodegradable scaffold; therefore, using sheet-like tissues, this technique allowed the facile fabrication of tubular tissues can be used as model. The fabricated artery tissue models had a multilayer structure. The assembly technique and perfusion culture system were applicable to many different sizes of fabricated arteries. The shape of the fabricated artery tissue models was maintained by the perfusion culture system; furthermore, the system reproduced the in vivo environment and allowed mechanical stimulation of the arteries. The multilayer structure of the artery tissue model was observed using fluorescent dyes. The equivalent Young's modulus was measured by applying internal pressure to the multilayer tubular tissues. The aim of this study was to determine whether fabricated artery tissue models maintained their mechanical properties with developing. We demonstrated both the rapid fabrication of multilayer tubular tissues that can be used as model arteries and the measurement of their equivalent Young's modulus in a suitable perfusion culture environment.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
PKH26 Linker-Kit für rot fluoreszierende Zellen für die allgemeine Zellmembranmarkierung, Distributed for Phanos Technologies
Sigma-Aldrich
PKH67 grün fluoreszierendes Zelllinker-Minikit für die allgemeine Zellmembranmarkierung, Distributed for Phanos Technologies
Sigma-Aldrich
PKH67 grün fluoreszierendes Zelllinker-Midikit für die allgemeine Zellmembranmarkierung, Distributed for Phanos Technologies