Merck
  • Listeria monocytogenes inhibition in refrigerated vacuum packaged turkey bologna by chemical additives.
<I>Listeria monocytogenes</I> inhibition in refrigerated vacuum packaged turkey bologna by chemical additives.
Journal of Food Science (1994)
Wederquist HJ, et al.