Merck
  • Preparation of photocatalytic TiO2 transparent thin film by thermal decomposition of Ti-alkoxide with a-terpineol as a solvent.
Preparation of photocatalytic TiO<SUB>2</SUB> transparent thin film by thermal decomposition of Ti-alkoxide with a-terpineol as a solvent.
Thin Solid Films (2001)
Negishi N and Takeuchi K.