Merck
  • Oligonucleotides designed to inhibit TLR9 block Herpes simplex virus type 1 infection at multiple steps.
Oligonucleotides designed to inhibit TLR9 block Herpes simplex virus type 1 infection at multiple steps.
Antiviral research (2014-07-06)
Monica M Sauter, Joshua J L Gauger, Curtis R Brandt
ZUSAMMENFASSUNG
Herpes simplex virus type 1 (HSV-1) is an important human pathogen which requires activation of nuclear factor-kappa B (NFκB) during its replication cycle. The persistent nature of HSV-1 infection, and the emergence of drug-resistant strains, highlights the importance of research to develop new antiviral agents. Toll-like receptors (TLRs) play a prominent role during the early antiviral response by recognizing viral nucleic acid and gene products, activating NFκB, and stimulating the production of inflammatory cytokines. We demonstrate a significant effect on HSV-1 replication in ARPE-19 and Vero cells when oligonucleotides designed to inhibit TLR9 are added 2h prior to infection. A greater than 90% reduction in the yield of infectious virus was achieved at oligonucleotide concentrations of 10-20 μM. TLR9 inhibitory oligonucleotides prevented expression of essential immediate early herpes gene products as determined by immunofluorescence microscopy and Western blotting. TLR9 oligonucleotides also interfered with viral attachment and entry. A TLR9 inhibitory oligonucleotide containing five adjacent guanosine residues (G-ODN) exhibited virucidal activity and inhibited HSV-1 replication when added post-infection. The antiviral effect of the TLR9 inhibitory oligonucleotides did not depend on the presence of TLR9 protein, suggesting a mechanism of inhibition that is not TLR9 specific. TLR9 inhibitory oligonucleotides also reduced NFκB activity in nuclear extracts. Studies using these TLR inhibitors in the context of viral infection should be interpreted with caution.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Proteasehemmer-Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Anti-Maus-IgG (Gesamtmolekül)−Alkalische Phosphatase in Ziege hergestellte Antikörper, affinity isolated antibody, buffered aqueous glycerol solution
Supelco
Acyclovir, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
NuCLEAR Extraction Kit, For mammalian tissue or cultured cells
USP
Acyclovir, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acycloguanosin, ≥99% (HPLC), powder
Aciclovir, European Pharmacopoeia (EP) Reference Standard