Merck
  • Overexpression of mitofilin in the mouse heart promotes cardiac hypertrophy in response to hypertrophic stimuli.
Overexpression of mitofilin in the mouse heart promotes cardiac hypertrophy in response to hypertrophic stimuli.
Antioxidants & redox signaling (2014-02-22)
Yuan Zhang, Jing Xu, Yu-Xuan Luo, Xi-Zhou An, Ran Zhang, Guang Liu, Hongliang Li, Hou-Zao Chen, De-Pei Liu
ZUSAMMENFASSUNG
Mitofilin was originally described as a heart muscle protein because of its abundance in the heart tissue; however, its function in the heart is still to be elucidated. Thus, this study aims at investigating the role of mitofilin in the heart in response to hypertrophic stimuli. In this study, a significant increase in mitofilin expression was observed in the hearts of patients with hypertrophic cardiomyopathy. Transgenic (TG) mice with cardiomyocyte-specific overexpression of mitofilin were generated, and cardiac hypertrophy was introduced by transverse aortic constriction (TAC) or chronic infusion of isoproterenol (ISO). In TG mice overexpressing mitofilin, the level of cardiac hypertrophy was significantly greater than that in wild-type (WT) mice after TAC and ISO stimulation. A detailed analysis showed that compared with WT mice, the level of reactive oxygen species was increased after TAC and ISO induction and mitochondrial oxidative phosphorylation (OXPHOS) activity in the TG hearts was lower. These alterations may contribute to the aggravated cardiac hypertrophy observed in response to TAC and ISO stimulation. Over-expression of mitofilin promotes cardiac hypertrophy under pathological conditions both in vivo and in vitro. Mitofilin, a mitochondria protein, is shown to be related to cardiac hypertrophy for the first time, which enhances our understanding of the role of mitochondria in cardiac hypertrophy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Ascorbinsäure, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbinsäure, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbinsäure, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbinsäure, 99%
Sigma-Aldrich
L-Ascorbinsäure, reagent grade, crystalline
Supelco
Ascorbinsäure, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Ascorbinsäure, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
L-Ascorbinsäure, ACS reagent, ≥99%
Supelco
L-Ascorbinsäure, analytical standard
Sigma-Aldrich
L-Ascorbinsäure, meets USP testing specifications
Sigma-Aldrich
Catalase Assay Kit, sufficient for ≥100 tests enzymatic, determination of catalase activity in tissues and cells
Sigma-Aldrich
bisBenzimid H 33342, ≥98% (HPLC)
Sigma-Aldrich
L-Ascorbinsäure, reagent grade
Sigma-Aldrich
DL-Isoproterenol -hydrochlorid
Sigma-Aldrich
L-Ascorbinsäure, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbinsäure, FCC, FG
Sigma-Aldrich
L-Ascorbinsäure, BioUltra, ≥99.5% (RT)
Ascorbinsäure, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Ascorbinsäure, puriss. p.a., ≥99.0% (RT)
Supelco
L-Ascorbinsäure, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Ascorbinsäure, tested according to Ph. Eur.