Merck
  • Unidirectional photoreceptor-to-Muller glia coupling and unique K+ channel expression in Caiman retina.
Unidirectional photoreceptor-to-Muller glia coupling and unique K+ channel expression in Caiman retina.
PLoS ONE (2014)
Zayas-Santiago A, Agte S, Rivera Y, et al.