Merck
  • One-tenth dose of LC 50 of 4-tert-butylphenol causes endocrine disruption and metabolic changes in Cyprinus carpio.
One-tenth dose of LC 50 of 4-tert-butylphenol causes endocrine disruption and metabolic changes in <I>Cyprinus carpio</I>.
Pesticide Biochemistry and Physiology (2006)
Barse AV, et al.