Merck
  • The relationship between taste and primary structure of ?delicious peptide?(Lys-Gly-Asp-Glu-Glu-Ser-Leu-Ala) from beef soup.
The relationship between taste and primary structure of ?delicious peptide?(Lys-Gly-Asp-Glu-Glu-Ser-Leu-Ala) from beef soup.
Agricultural and Biological Chemistry (1989)
Tamura M, et al.