Merck
  • Bis (trifluoromethane) sulfonimide initiated ring-opening polymerization of octamethylcyclotetrasiloxane.
Bis (trifluoromethane) sulfonimide initiated ring-opening polymerization of octamethylcyclotetrasiloxane.
Journal of Organometallic Chemistry (2002)
Desmurs J-R, et al.