Merck
  • Linkage of inflammation and oxidative stress via release of glutathionylated peroxiredoxin-2, which acts as a danger signal.
Linkage of inflammation and oxidative stress via release of glutathionylated peroxiredoxin-2, which acts as a danger signal.
Proceedings of the National Academy of Sciences of the USA (2014)
Salzano S, Checconi P, Hanschmann EM, et al.